gRNA Design Details
- Target DNA Sequence
- AGTCGCTGTTGGGTCTCATG
- PAM Sequence
- GGG
- Target locus
-
Chr.5: 38451396 - 38451418 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EGFLAM
- Entrez Gene ID
-
133584
- Gene Name
- EGF like, fibronectin type III and laminin G domains
- Gene Aliases
-
AGRINL, AGRNL, PIKA
- Chromosome Location
-
Chr.5: 38258409 - 38465480 on GRCh38
- UniGene ID
- Hs.20103
- Species
-
Human