gRNA Design Details
- Target DNA Sequence
- TGGACACGGTGAAGAAGTAG
- PAM Sequence
- TGG
- Target locus
-
Chr.18: 77250881 - 77250903 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GALR1
- Entrez Gene ID
-
2587
- Gene Name
- galanin receptor 1
- Gene Aliases
-
GALNR, GALNR1
- Chromosome Location
-
Chr.18: 77250052 - 77270142 on GRCh38
- UniGene ID
- Hs.272191
- Species
-
Human