gRNA Design Details
- Target DNA Sequence
- AGAGTCCGGCTGTTGTATTT
- PAM Sequence
- GGG
- Target locus
-
Chr.7: 142773354 - 142773376 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.63/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- PRSS2
- Entrez Gene ID
-
5645
- Gene Name
- protease, serine 2
- Gene Aliases
-
TRY2, TRY8, TRYP2
- Chromosome Location
-
Chr.7: 142770943 - 142774564 on GRCh38
- UniGene ID
- Hs.449281
- Species
-
Human