gRNA Design Details

Target DNA Sequence
CTCTCATGTCGAGCTAGCAG
PAM Sequence
TGG
Target locus
Chr.14: 74683449 - 74683471 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.89/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AREL1
Entrez Gene ID
9870
Gene Name
apoptosis resistant E3 ubiquitin protein ligase 1
Gene Aliases
FIEL1, KIAA0317
Chromosome Location
Chr.14: 74661252 - 74713118 on GRCh38
UniGene ID
Hs.497417
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001039479.1 NP_001034568.1 5
XM_005268244.3 XP_005268301.1 4
XM_006720344.2 XP_006720407.1 5
XM_011537415.2 XP_011535717.1 6
XM_011537416.2 XP_011535718.1 5
XM_011537417.2 XP_011535719.1 5