gRNA Design Details
- Target DNA Sequence
- CGCAAGTCAGGAAGAAGCGC
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 43304900 - 43304922 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TIE1
- Entrez Gene ID
-
7075
- Gene Name
- tyrosine kinase with immunoglobulin like and EGF like domains 1
- Gene Aliases
-
JTK14, TIE
- Chromosome Location
-
Chr.1: 43300895 - 43323110 on GRCh38
- UniGene ID
- Hs.78824
- Species
-
Human