gRNA Design Details
- Target DNA Sequence
- GCGATGCTGGACCAGTTGAT
- PAM Sequence
- GGG
- Target locus
-
Chr.7: 139360286 - 139360308 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- LUC7L2
- Entrez Gene ID
-
51631
- Gene Name
- LUC7 like 2, pre-mRNA splicing factor
- Gene Aliases
-
CGI-59, CGI-74, LUC7B2
- Chromosome Location
-
Chr.7: 139340359 - 139423457 on GRCh38
- UniGene ID
- Hs.731488
- Species
-
Human