gRNA Design Details

Target DNA Sequence
TGTAGAAGAGTTTCCAGCCG
PAM Sequence
CGG
Target locus
Chr.15: 98707785 - 98707807 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 88.96/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
IGF1R
Entrez Gene ID
3480
Gene Name
insulin like growth factor 1 receptor
Gene Aliases
CD221, IGFIR, IGFR, JTK13
Chromosome Location
Chr.15: 98648539 - 98964530 on GRCh38
UniGene ID
Hs.643120
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000875.4 NP_000866.1 2
NM_001291858.1 NP_001278787.1 2
XM_017022136.1 XP_016877625.1 2
XM_017022137.1 XP_016877626.1 2
XM_017022138.1 XP_016877627.1 2
XM_017022139.1 XP_016877628.1 2