gRNA Design Details
- Target DNA Sequence
- GAAAGTCCGTGACTTGCAAG
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 140345932 - 140345954 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
90.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HBEGF
- Entrez Gene ID
-
1839
- Gene Name
- heparin binding EGF like growth factor
- Gene Aliases
-
DTR, DTS, DTSF, HEGFL
- Chromosome Location
-
Chr.5: 140332843 - 140346603 on GRCh38
- UniGene ID
- Hs.592942
- Species
-
Human