gRNA Design Details

Target DNA Sequence
TCGTCTGTCGGACACGTTGA
PAM Sequence
AGG
Target locus
Chr.6: 27248014 - 27248036 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 95.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS16
Entrez Gene ID
10279
Gene Name
protease, serine 16
Gene Aliases
TSSP
Chromosome Location
Chr.6: 27247701 - 27256620 on GRCh38
UniGene ID
Hs.274407
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_005865.3 NP_005856.1 2
XM_017010161.1 XP_016865650.1 2
XM_017010162.1 XP_016865651.1 2
XM_017010163.1 XP_016865652.1 2
XM_017010164.1 XP_016865653.1 2
XM_017010165.1 XP_016865654.1 2