gRNA Design Details
- Target DNA Sequence
- CACCGACGTGTAGCAGCAGA
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 100473600 - 100473622 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.95/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TMEFF1
- Entrez Gene ID
-
8577
- Gene Name
- transmembrane protein with EGF like and two follistatin like domains 1
- Gene Aliases
-
C9orf2, CT120.1, H7365, TR-1
- Chromosome Location
-
Chr.9: 100473238 - 100577636 on GRCh38
- UniGene ID
- Hs.598100
- Species
-
Human