gRNA Design Details
- Target DNA Sequence
- GATGGAAGCCTGCTGGAAAC
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 96160609 - 96160631 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF6
- Entrez Gene ID
-
392255
- Gene Name
- growth differentiation factor 6
- Gene Aliases
-
BMP-13, BMP13, CDMP2, KFM, KFS, KFS1, KFSL, LCA17, MCOP4, MCOPCB6, SCDO4, SGM1
- Chromosome Location
-
Chr.8: 96142330 - 96160792 on GRCh38
- UniGene ID
- Hs.492277
- Species
-
Human