gRNA Design Details
- Target DNA Sequence
- ATGATGCTTGCCGGAGCCTG
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 64284255 - 64284277 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
78.6/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- BAD
- Entrez Gene ID
-
572
- Gene Name
- BCL2 associated agonist of cell death
- Gene Aliases
-
BBC2, BCL2L8
- Chromosome Location
-
Chr.11: 64269828 - 64284704 on GRCh38
- UniGene ID
- Hs.370254
- Species
-
Human