gRNA Design Details
- Target DNA Sequence
- GTCCGTGGGGATGGACAACT
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 113981255 - 113981277 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.34/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HTR3A
- Entrez Gene ID
-
3359
- Gene Name
- 5-hydroxytryptamine receptor 3A
- Gene Aliases
-
5-HT-3, 5-HT3A, 5-HT3R, 5HT3R, HTR3
- Chromosome Location
-
Chr.11: 113975075 - 113990313 on GRCh38
- UniGene ID
- Hs.413899
- Species
-
Human