gRNA Design Details
- Target DNA Sequence
- TGGGAACTATACAGGGAGCT
- PAM Sequence
- GGG
- Target locus
-
Chr.11: 124625784 - 124625806 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TBRG1
- Entrez Gene ID
-
84897
- Gene Name
- transforming growth factor beta regulator 1
- Gene Aliases
-
NIAM, TB-5
- Chromosome Location
-
Chr.11: 124622599 - 124635926 on GRCh38
- UniGene ID
- Hs.436410
- Species
-
Human