gRNA Design Details
- Target DNA Sequence
- ATGGTGAACTCGCTGCTGTT
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 18956310 - 18956332 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IFFO2
- Entrez Gene ID
-
126917
- Gene Name
- intermediate filament family orphan 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 18904280 - 18956672 on GRCh38
- UniGene ID
- Hs.466625
- Species
-
Human