gRNA Design Details
- Target DNA Sequence
- TACAGCAGTCGGAGAACATC
- PAM Sequence
- TGG
- Target locus
-
Chr.20: 43459785 - 43459807 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
79.89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRSF6
- Entrez Gene ID
-
6431
- Gene Name
- serine and arginine rich splicing factor 6
- Gene Aliases
-
B52, HEL-S-91, SFRS6, SRP55
- Chromosome Location
-
Chr.20: 43457864 - 43463605 on GRCh38
- UniGene ID
- Hs.684950
- Species
-
Human