Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001287593.1 | NP_001274522.1 | 3 | 696 | |
NM_001287594.1 | NP_001274523.1 | 3 | 236 | ||
NM_001288.4 | NP_001279.2 | 2 | 387 | ||
GenBank mRNA | AF034607.1 | - | 2 | 392 | |
AF109197.1 | - | 2 | 177 | ||
AK129773.1 | - | 2 | 132 | ||
AK223379.1 | - | 2 | 360 | ||
AK314014.1 | - | 2 | 132 | ||
BC064527.1 | - | 3 | 696 | 5585323 | |
BC095469.1 | - | 2 | 322 | 30557606 | |
BM562536.1 | - | 390 | |||
CR542071.1 | - | 2 | 123 | ||
CX783058.1 | - | 3 | 234 | ||
HY057217.1 | - | 3 | 237 | ||
U93205.1 | - | 2 | 341 | ||
X87689.1 | - | 2 | 327 |