Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164277.1 | NP_001157749.1 | 6 | 1453 | |
NM_001164278.1 | NP_001157750.1 | 6 | 1137 | ||
NM_001164279.1 | NP_001157751.1 | 6 | 879 | ||
NM_001164280.1 | NP_001157752.1 | 4 | 895 | ||
NM_001467.5 | NP_001458.1 | 5 | 949 | ||
GenBank mRNA | AB209026.1 | - | 2 | 3306 | |
AF110819.1 | - | 4 | 696 | ||
AF110820.1 | - | 4 | 696 | ||
AF110821.1 | - | 3 | 1017 | ||
AF110822.1 | - | 4 | 696 | ||
AF111852.1 | - | 2 | 1449 | ||
AK289642.1 | - | 6 | 1421 | ||
AK294993.1 | - | 6 | 1015 | ||
AK298395.1 | - | 6 | 833 | ||
AK300646.1 | - | 6 | 845 | ||
AK312628.1 | - | 6 | 1119 | ||
AY423732.1 | - | 4 | 716 | ||
BC002400.2 | - | 4 | 897 | 2822215 | |
BC003589.1 | - | 4 | 897 | 2822215 | |
BC014663.1 | - | 4 | 904 | 4857067 | |
BC015650.2 | - | 5 | 1369 | 4660508 | |
BC064563.1 | - | 4 | 892 | 5580134 | |
CR456733.1 | - | 4 | 696 | ||
Y15409.1 | - | 4 | 865 |