Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193508.1 | NP_001180437.1 | 2 | 879 | |
NM_005612.4 | NP_005603.3 | 2 | 1090 | ||
XM_011534401.2 | XP_011532703.1 | 2 | 1197 | ||
XM_017008527.1 | XP_016864016.1 | 2 | 1051 | ||
GenBank mRNA | AB209750.1 | - | 2 | 830 | |
AF228045.1 | - | 1 | 170 | ||
BC017822.1 | - | 2 | 781 | ||
BC038985.1 | - | 2 | 1019 | ||
BC132859.1 | - | 2 | 766 | 40146881 | |
BC136491.1 | - | 2 | 766 | 9020480 | |
JX896957.1 | - | 2 | 872 | ||
JX896958.1 | - | 2 | 872 | ||
JX896959.1 | - | 2 | 872 | ||
JX896966.1 | - | 2 | 776 | ||
JX896971.1 | - | 2 | 872 | ||
JX896975.1 | - | 3 | 187 | ||
JX896981.1 | - | 2 | 776 | ||
JX896983.1 | - | 2 | 872 | ||
JX896984.1 | - | 2 | 872 | ||
JX896989.1 | - | 2 | 776 | ||
JX896990.1 | - | 2 | 779 | ||
JX896991.1 | - | 2 | 872 | ||
JX896992.1 | - | 2 | 776 | ||
JX896993.1 | - | 2 | 779 | ||
KC117266.1 | - | 3 | 476 | ||
U13877.1 | - | 1 | 753 | ||
U13879.1 | - | 2 | 990 | ||
U22314.1 | - | 1 | 792 |