Gene Transcripts

Gene Symbol
WEE1
Entrez Gene ID
7465
Gene Name
WEE1 G2 checkpoint kinase
Gene Aliases
WEE1A, WEE1hu
Chromosome Location
Chr. 11: 9573681 - 9589767 on Build GRCh38
UniGene ID
Hs.249441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143976.1 NP_001137448.1 5 1220
NM_003390.3 NP_003381.1 5 1360
GenBank mRNA AK122837.1 - 5 1220
BC051831.1 - 5 1158 6147695
BC070052.1 - 5 1350 30336707
BX641032.1 - 4 878
U10564.1 - 5 1360
X62048.1 - 5 1277

View Full Product Details