Gene Transcripts

Gene Symbol
HSD17B8
Entrez Gene ID
7923
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases
D6S2245E, FABG, FABGL, H2-KE6, HKE6, KE6, RING2, SDR30C1, dJ1033B10.9
Chromosome Location
Chr. 6: 33204637 - 33206831 on Build GRCh38
UniGene ID
Hs.415058
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014234.4 NP_055049.1 8 763
GenBank mRNA BC008185.1 - 8 758
BT007239.1 - 8 731
D82061.1 - 8 727

View Full Product Details