Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001178010.2 | NP_001171481.1 | 14 | 1372 | |
NM_001178011.2 | NP_001171482.1 | 12 | 1138 | ||
NM_003504.4 | NP_003495.1 | 13 | 1276 | ||
XM_005261285.2 | XP_005261342.1 | 15 | 1465 | ||
XM_005261286.3 | XP_005261343.1 | 14 | 1369 | ||
XM_011530415.1 | XP_011528717.1 | 14 | 1369 | ||
XM_011530416.1 | XP_011528718.1 | 13 | 1172 | ||
XM_011530417.2 | XP_011528719.1 | 13 | 1432 | ||
XM_011530418.2 | XP_011528720.1 | 13 | 1432 | ||
XM_017028966.1 | XP_016884455.1 | 8 | 751 | ||
XM_017028967.1 | XP_016884456.1 | 5 | 491 | ||
GenBank mRNA | AF053074.1 | - | 13 | 1157 | |
AF062495.1 | - | 13 | 1149 | ||
AF081535.1 | - | 13 | 1164 | ||
AJ223728.1 | - | 13 | 1163 | ||
AK293123.1 | - | 12 | 1073 | ||
AK293338.1 | - | 14 | 1269 | ||
AK303274.1 | - | 13 | 1196 | ||
AY358971.1 | - | 13 | 1199 | ||
BC006232.2 | - | 13 | 1194 | 3946493 | |
BC010022.2 | - | 13 | 1179 | 2964592 | |
BT006792.1 | - | 13 | 1133 | ||
CT841513.1 | - | 13 | 1137 |