Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278720.1 | NP_001265649.1 | 8 | 1190 | |
NM_001278721.1 | NP_001265650.1 | 7 | 965 | ||
NM_001318733.1 | NP_001305662.1 | 7 | 1287 | ||
XM_017023849.1 | XP_016879338.1 | 6 | 1189 | ||
XM_017023850.1 | XP_016879339.1 | 5 | 1270 | ||
GenBank mRNA | AJ272344.1 | - | 8 | 1150 | |
AK294136.1 | - | 6 | 1294 | ||
BC120874.2 | - | 7 | 1279 | 40081041 | |
BC120875.2 | - | 7 | 1280 | 40081042 | |
BM674847.1 | - | 1 | 287 | ||
Y17108.1 | - | 7 | 1287 |