Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010938.1 | NP_001010938.1 | 6 | 1170 | |
NM_001308046.1 | NP_001294975.1 | 6 | 1228 | ||
NM_005781.4 | NP_005772.3 | 6 | 1379 | ||
XM_005269268.3 | XP_005269325.1 | 6 | 1170 | ||
XM_005269270.3 | XP_005269327.1 | 6 | 1018 | ||
XM_005269274.3 | XP_005269331.1 | 3 | 435 | ||
XM_011512317.2 | XP_011510619.1 | 8 | 1737 | ||
XM_011512318.2 | XP_011510620.2 | 6 | 1109 | ||
XM_011512320.1 | XP_011510622.1 | 6 | 892 | ||
XM_011512321.2 | XP_011510623.1 | 4 | 709 | ||
XM_017005508.1 | XP_016860997.1 | 6 | 1109 | ||
XM_017005509.1 | XP_016860998.1 | 6 | 1109 | ||
XM_017005510.1 | XP_016860999.1 | 6 | 1109 | ||
GenBank mRNA | AB209338.1 | - | 3 | 3237 | |
AK131539.1 | - | 6 | 1170 | ||
AK225786.1 | - | 6 | 1175 | ||
BC008884.2 | - | 6 | 1025 | ||
BC028164.1 | - | 6 | 893 | ||
L13738.2 | - | 6 | 1454 |