Gene Transcripts
- Gene Symbol
- TMC6
- Entrez Gene ID
- 11322
- Gene Name
- transmembrane channel like 6
- Gene Aliases
- EV1, EVER1, EVIN1, LAK-4P
- Chromosome Location
- Chr. 17: 78108775 - 78132427 on Build GRCh38
- UniGene ID
- Hs.632227
- Species
- Homo sapiens
- Gene Symbol
- LOC339539
- Entrez Gene ID
- 339539
- Gene Name
- uncharacterized LOC339539
- Gene Aliases
- Chromosome Location
- Chr. 1: 42857622 - 42888792 on Build GRCh38
- UniGene ID
- Hs.434301
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104171.1 |
|
3, 4 |
255 |
|
GenBank mRNA |
BC043541.1 |
- |
3, 4 |
255 |
|
View Full Product Details