Gene Transcripts

Gene Symbol
IRX4
Entrez Gene ID
50805
Gene Name
iroquois homeobox 4
Gene Aliases
IRXA3
Chromosome Location
Chr. 5: 1877427 - 1887179 on Build GRCh38
UniGene ID
Hs.196927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278632.1 NP_001265561.1 4, 5 626
NM_001278634.1 NP_001265563.1 4, 5 631
NM_016358.2 NP_057442.1 3, 4 527
GenBank mRNA AB690778.1 - 4, 5 626
AB690780.1 - 4, 5 631
AF124733.1 - 3, 4 527
BC110912.1 - 3, 4 415 3450161
BC136505.1 - 3, 4 515 9020494
BC136506.1 - 3, 4 515 9020495
U90306.1 - 2, 3 172

View Full Product Details