Gene Transcripts

Gene Symbol
SPG21
Entrez Gene ID
51324
Gene Name
spastic paraplegia 21 (autosomal recessive, Mast syndrome)
Gene Aliases
ACP33, BM-019, GL010, MAST
Chromosome Location
Chr. 15: 64963021 - 64989946 on Build GRCh38
UniGene ID
Hs.242458
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127889.4 NP_001121361.1 3 254
NM_001127890.4 NP_001121362.1 3 435
NM_016630.6 NP_057714.1 3 435
XM_005254437.4 XP_005254494.1 4 468
XM_017022297.1 XP_016877786.1 3 803
XM_017022298.1 XP_016877787.1 4 893
GenBank mRNA AF208861.1 - 2 150
AF212231.1 - 3 222
AK123725.1 - 1 667
AK172849.1 - 3 267
AK225527.1 - 3 267
AK312943.1 - 3 269
BC000244.1 - 3 305 3357388
BI552731.1 - 3 405
BI554234.1 - 4 463
BI771887.1 - 3 269
BP268459.1 - 3 428
BP268875.1 - 3 428
BX446863.2 - 3 385
DA680695.1 - 3 253

View Full Product Details