Gene Transcripts

Gene Symbol
ANAPC5
Entrez Gene ID
51433
Gene Name
anaphase promoting complex subunit 5
Gene Aliases
APC5
Chromosome Location
Chr. 12: 121308245 - 121354209 on Build GRCh38
UniGene ID
Hs.7101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137559.1 NP_001131031.1 17 2114
NM_016237.4 NP_057321.2 17 2375
XM_005253900.3 XP_005253957.1 17 2338
XM_017019423.1 XP_016874912.1 16 2011
XM_017019424.1 XP_016874913.1 16 1972
GenBank mRNA AF191339.1 - 17 2253
AK025614.1 - 17 2114
AK054779.1 - 7 1951
AK294139.1 - 10 1371
AK301197.1 - 9 1605
AK315451.1 - 17 2320
BC001081.2 - 17 2308 2823401
BC001950.1 - 17 2308 2823401
BC003095.1 - 12 1577
BC006301.2 - 17 2312 4100632
BC034243.1 - 16 2015
BC045686.1 - 10 2463
BX381618.2 - 2 258
BX537687.1 - 12 2002

View Full Product Details