Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001288607.1 | NP_001275536.1 | 10 | 1862 | |
NM_001288608.1 | NP_001275537.1 | 10 | 1862 | ||
NM_019032.5 | NP_061905.2 | 9 | 1793 | ||
NM_025008.4 | NP_079284.2 | 9 | 1793 | ||
XM_011509644.2 | XP_011507946.1 | 9 | 1824 | ||
XM_011509645.2 | XP_011507947.1 | 8 | 1752 | ||
XM_011509648.2 | XP_011507950.1 | 9 | 1778 | ||
XM_011509649.2 | XP_011507951.1 | 9 | 1832 | ||
XM_011509650.2 | XP_011507952.1 | 9 | 1832 | ||
XM_011509651.2 | XP_011507953.1 | 3 | 574 | ||
XM_011509652.1 | XP_011507954.1 | 3 | 280 | ||
XM_017001506.1 | XP_016856995.1 | 10 | 2233 | ||
XM_017001507.1 | XP_016856996.1 | 2 | 188 | ||
GenBank mRNA | AF370411.1 | - | 3 | 328 | |
AY358122.1 | - | 8 | 1607 | ||
BC027478.1 | - | 2 | 232 | ||
BC071852.1 | - | 1 | 89 | ||
BC094811.1 | - | 1 | 89 | ||
BC140800.1 | - | 9 | 1714 | 9021668 | |
BC144581.1 | - | 9 | 1714 | 9053111 | |
BX282114.1 | - | 3 | 398 |