Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020208.3 | NP_064593.1 | 2 | 271 | |
NM_022405.3 | NP_071800.1 | 2 | 271 | ||
XM_011533848.2 | XP_011532150.1 | 2 | 281 | ||
GenBank mRNA | AF075260.1 | - | 2 | 149 | |
AF125107.1 | - | 3 | 335 | ||
AJ276207.1 | - | 2 | 198 | ||
AJ276208.1 | - | 2 | 198 | ||
BC126197.1 | - | 2 | 211 | 8991913 | |
BC136431.1 | - | 2 | 211 | 9020420 | |
BC144087.1 | - | 2 | 211 | 9052607 | |
DR006419.1 | - | 2 | 278 |