Gene Transcripts

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
Chromosome Location
Chr. 11: 44066118 - 44084237 on Build GRCh38
UniGene ID
Hs.126706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127219.1 NP_001120691.1 13, 14 1386
NM_032592.3 NP_115981.1 13, 14 1682
XM_005253170.3 XP_005253227.2 13, 14 1825
XM_005253172.2 XP_005253229.1 10, 11 913
XM_006718348.2 XP_006718411.1 13, 14 1134
XM_011520407.1 XP_011518709.1 13, 14 1178
XM_011520410.2 XP_011518712.1 13, 14 2079
GenBank mRNA AK057649.1 - 13, 14 1676
AL833533.1 - 5, 6 2679
AY026508.1 - 13, 14 1637
BC020197.1 - 13, 14 1656 3632065
KU178811.1 - 12, 13 1242
KU178812.1 - 12, 13 1242
KU178813.1 - 11, 12 1182

View Full Product Details