Gene Transcripts

Gene Symbol
C11orf40
Entrez Gene ID
143501
Gene Name
chromosome 11 open reading frame 40
Gene Aliases
NOV1
Chromosome Location
Chr. 11: 4571423 - 4577820 on Build GRCh38
UniGene ID
Hs.350556
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144663.1 NP_653264.1 4 613
GenBank mRNA AF439154.1 - 4 613

View Full Product Details