Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152431.2 | NP_689644.2 | 2 | 342 | |
GenBank mRNA | AB079366.1 | - | 2 | 302 | |
AK093475.1 | - | 2 | 282 | ||
AK293616.1 | - | 2 | 194 | ||
BC031060.1 | - | 2 | 134 | 5271278 | |
CR749642.1 | - | 2 | 270 | ||
DB091271.1 | - | 2 | 342 |