Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173674.2 | NP_775945.1 | 9, 10 | 1289 | |
XM_005266941.1 | XP_005266998.1 | 9, 10 | 1253 | ||
XM_006715463.1 | XP_006715526.1 | 9, 10 | 1253 | ||
XM_011535771.1 | XP_011534073.1 | 10, 11 | 1338 | ||
XM_011535773.2 | XP_011534075.1 | 9, 10 | 1253 | ||
XM_011535774.2 | XP_011534076.1 | 9, 10 | 1253 | ||
XM_017010775.1 | XP_016866264.1 | 10, 11 | 1262 | ||
GenBank mRNA | AK095973.1 | - | 9, 10 | 1159 | |
BC035671.1 | - | 9, 10 | 1236 | 5730536 | |
CR749448.1 | - | 6 | 2841 |