Gene Transcripts

Gene Symbol
MYLK
Entrez Gene ID
4638
Gene Name
myosin light chain kinase
Gene Aliases
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
Chromosome Location
Chr. 3: 123612296 - 123884302 on Build GRCh38
UniGene ID
Hs.477375
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321309.1 NP_001308238.1 33 5636
NM_053025.3 NP_444253.3 34 5844
NM_053026.3 NP_444254.3 33 5637
NM_053027.3 NP_444255.3 33 5691
NM_053028.3 NP_444256.3 32 5484
XM_011512860.2 XP_011511162.1 31 5562
XM_011512861.2 XP_011511163.1 30 5361
XM_017006469.1 XP_016861958.1 17 3120
XM_017006470.1 XP_016861959.1 16 2108
XM_017006471.1 XP_016861960.1 16 2161
XM_017006472.1 XP_016861961.1 4 454
XM_017006473.1 XP_016861962.1 4 403
GenBank mRNA AB037663.1 - 17 2920
AF069601.2 - 31 5474
AF069602.1 - 31 5525
AF069603.1 - 30 5318
AF069604.1 - 11 1500
AF096773.1 - 3 478
AF096774.1 - 3 475
AF096775.1 - 3 478
AK127437.1 - 1 1367
AK300610.1 - 16 2094
AK314412.1 - 3 489
AK314443.1 - 3 532
AY339601.1 - 31 5562
AY424269.1 - 30 5357
AY424270.1 - 31 5564
BC017811.1 - 3 499
BC064420.1 - 3 518
BC100761.2 - 3 360
BC100762.1 - 3 360
BC100763.1 - 3 357
BC107783.1 - 3 502
BC113456.1 - 3 323
BC113458.1 - 3 320
CR749364.1 - 3 5304
U48959.2 - 32 5681
X85337.1 - 18 3174
X90870.1 - 9 1194

View Full Product Details