Gene Transcripts

Gene Symbol
HPR
Entrez Gene ID
3250
Gene Name
haptoglobin-related protein
Gene Aliases
A-259H10.2, HP
Chromosome Location
Chr. 16: 72063226 - 72077246 on Build GRCh38
UniGene ID
Hs.655361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020995.3 NP_066275.3 3 161
GenBank mRNA AV698541.1 - 3 146
CB115435.1 - 3 160
CB147217.1 - 3 161
T71114.1 - 3 146

View Full Product Details