Gene Transcripts

Gene Symbol
PMP22
Entrez Gene ID
5376
Gene Name
peripheral myelin protein 22
Gene Aliases
CMT1A, CMT1E, DSS, GAS-3, GAS3, HMSNIA, HNPP, Sp110
Chromosome Location
Chr. 17: 15229777 - 15265357 on Build GRCh38
UniGene ID
Hs.372031
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000304.3 NP_000295.1 5 612
NM_001281455.1 NP_001268384.1 5 573
NM_001281456.1 NP_001268385.1 5 608
NM_153321.2 NP_696996.1 5 569
NM_153322.2 NP_696997.1 4 491
NR_104017.1 4 500
NR_104018.1 3 400
GenBank mRNA AK290640.1 - 5 522
AK300690.1 - 5 464
BC019040.2 - 5 534 4588473
BC091499.1 - 5 529 30395315
BQ232542.1 - 3 281
BX464118.2 - 5 567
CD519144.1 - 1 66
CR541931.1 - 4 374
CR541953.1 - 4 374
D11428.1 - 5 562
HY099893.1 - 3 370
KR259962.1 - 3 356
L03203.1 - 5 488
M94048.1 - 4 423
X65968.1 - 2 195

View Full Product Details