Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013943.2 | NP_039234.1 | 2 | 441 | |
GenBank mRNA | AF097330.1 | - | 2 | 296 | |
AF109196.1 | - | 2 | 234 | ||
AK001624.1 | - | 2 | 328 | ||
AK095959.1 | - | 2 | 262 | ||
AK301466.1 | - | 2 | 239 | ||
AK315051.1 | - | 2 | 278 | ||
AL117424.1 | - | 2 | 353 | ||
BC012444.1 | - | 2 | 306 | 3861372 | |
CR533501.1 | - | 2 | 156 | ||
DA904028.1 | - | 2 | 397 | ||
DB091421.1 | - | 2 | 444 |