Gene Transcripts

Gene Symbol
DISP3
Entrez Gene ID
57540
Gene Name
dispatched RND transporter family member 3
Gene Aliases
PTCHD2
Chromosome Location
Chr. 1: 11479155 - 11537583 on Build GRCh38
UniGene ID
Hs.202355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020780.1 NP_065831.1 3, 4 1444
XM_011541828.2 XP_011540130.1 2, 3 3546
XM_011541829.1 XP_011540131.1 2, 3 419
XM_011541830.2 XP_011540132.1 3, 4 1527
XM_011541831.2 XP_011540133.1 3, 4 1527
XM_011541832.2 XP_011540134.1 3, 4 1527
XM_011541833.2 XP_011540135.1 3, 4 1527
XM_011541835.2 XP_011540137.1 3, 4 1527
GenBank mRNA AB037758.1 - 3, 4 1444
AB593117.1 - 3, 4 1527
AL117236.1 - 2, 3 364

View Full Product Details