Gene Transcripts

Gene Symbol
FBXO41
Entrez Gene ID
150726
Gene Name
F-box protein 41
Gene Aliases
FBX41
Chromosome Location
Chr. 2: 73254682 - 73284453 on Build GRCh38
UniGene ID
Hs.23158
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080410.2 NP_001073879.2 9 2199
XM_005264158.3 XP_005264215.1 7 1118
XM_006711950.2 XP_006712013.1 14 3183
XM_011532562.1 XP_011530864.1 10 2631
XM_011532563.2 XP_011530865.1 8 1556
XM_017003421.1 XP_016858910.1 11 5280
XM_017003422.1 XP_016858911.1 14 4925
XM_017003423.1 XP_016858912.1 14 4306
GenBank mRNA AB075820.1 - 9 2037
AK127503.1 - 6 680
BC112202.1 - 8 1107

View Full Product Details