Gene Transcripts
- Gene Symbol
- LOC100289333
- Entrez Gene ID
- 100289333
- Gene Name
- uncharacterized LOC100289333
- Gene Aliases
- Chromosome Location
- Chr. 19: 12195015 - 12207574 on Build GRCh38
- UniGene ID
- Hs.662603
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104179.1 |
|
2 |
305 |
|
GenBank mRNA |
BX105180.1 |
- |
2 |
209 |
|
View Full Product Details