Gene Transcripts

Gene Symbol
C1orf137
Entrez Gene ID
388667
Gene Name
chromosome 1 open reading frame 137
Gene Aliases
Chromosome Location
Chr. 1: 116694112 - 116706603 on Build GRCh38
UniGene ID
Hs.127851
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013643.1 NP_001013665.1 4 300
GenBank mRNA AA922397.1 - 1 83

View Full Product Details