Gene Transcripts
- Gene Symbol
- LINC01568
- Entrez Gene ID
- 100506172
- Gene Name
- long intergenic non-protein coding RNA 1568
- Gene Aliases
- Chromosome Location
- Chr. 16: 73386805 - 73421396 on Build GRCh38
- UniGene ID
- Hs.434230
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042734.1 |
- |
2 |
330 |
|
View Full Product Details