Gene Transcripts
- Gene Symbol
- C19orf18
- Entrez Gene ID
- 147685
- Gene Name
- chromosome 19 open reading frame 18
- Gene Aliases
- Chromosome Location
- Chr. 19: 57958437 - 57974534 on Build GRCh38
- UniGene ID
- Hs.134209
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152474.4 |
NP_689687.1
|
1, 2 |
217 |
|
GenBank mRNA |
BC033933.2 |
- |
1, 2 |
220 |
|
View Full Product Details