Gene Transcripts

Gene Symbol
CIART
Entrez Gene ID
148523
Gene Name
circadian associated repressor of transcription
Gene Aliases
C1orf51, CHRONO, GM129
Chromosome Location
Chr. 1: 150282533 - 150287093 on Build GRCh38
UniGene ID
Hs.54680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300838.1 NP_001287767.1 3 557
NM_001300839.1 NP_001287768.1 3 303
NM_001300840.1 NP_001287769.1 3 251
NM_001300841.1 NP_001287770.1 3 319
NM_144697.3 NP_653298.1 2 1146
GenBank mRNA AK056960.1 - 2 427
AK315397.1 - 2 831
BC017397.1 - 3 275
BC027999.1 - 3 547 5194413
BC047238.1 - 2 860 5273646
BE784098.1 - 3 249
CN359562.1 - 3 217

View Full Product Details