Gene Transcripts

Gene Symbol
CCDC140
Entrez Gene ID
151278
Gene Name
coiled-coil domain containing 140
Gene Aliases
Chromosome Location
Chr. 2: 222298147 - 222305217 on Build GRCh38
UniGene ID
Hs.350729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153038.1 NP_694583.1 2 531
XM_017003466.1 XP_016858955.1 2 372
GenBank mRNA AK057009.1 - 2 531
BC109388.1 - 2 436 40023803

View Full Product Details