Gene Transcripts

Gene Symbol
JAZF1
Entrez Gene ID
221895
Gene Name
JAZF zinc finger 1
Gene Aliases
TIP27, ZNF802
Chromosome Location
Chr. 7: 27830573 - 28180818 on Build GRCh38
UniGene ID
Hs.368944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175061.3 NP_778231.2 4 770
XM_006715656.1 XP_006715719.1 5 646
XM_017011831.1 XP_016867320.1 4 538
XM_017011832.1 XP_016867321.1 4 470
GenBank mRNA AK091311.1 - 4 695
AL832491.1 - 2 348
AL834234.1 - 3 489
AY372319.1 - 4 548
BC042441.1 - 5 558 4831063
BC047229.1 - 4 557

View Full Product Details