Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015276.1 | NP_056091.1 | 11 | 1565 | |
XM_005256575.2 | XP_005256632.1 | 11 | 1279 | ||
GenBank mRNA | AB028986.1 | - | 11 | 1565 | |
BC007196.1 | - | 4 | 307 | ||
BC025317.1 | - | 4 | 305 | ||
BC110499.1 | - | 11 | 1324 | ||
BC126898.1 | - | 11 | 1325 | 40027759 | |
BX640815.1 | - | 10 | 1147 |