Gene Transcripts

Gene Symbol
GBX1
Entrez Gene ID
2636
Gene Name
gastrulation brain homeobox 1
Gene Aliases
Huh-17
Chromosome Location
Chr. 7: 151148009 - 151167548 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098834.2 NP_001092304.1 2 845
XM_017011964.1 XP_016867453.1 3 862
XM_017011965.1 XP_016867454.1 3 859

View Full Product Details