Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010986.2 | NP_001010986.1 | 12 | 1214 | |
NM_173694.4 | NP_775965.2 | 12 | 1214 | ||
XM_017029439.1 | XP_016884928.1 | 13 | 1220 | ||
XM_017029440.1 | XP_016884929.1 | 13 | 1891 | ||
XM_017029441.1 | XP_016884930.1 | 14 | 1300 | ||
XM_017029442.1 | XP_016884931.1 | 13 | 1441 | ||
XM_017029443.1 | XP_016884932.1 | 13 | 1219 | ||
XM_017029444.1 | XP_016884933.1 | 12 | 1831 | ||
XM_017029445.1 | XP_016884934.1 | 13 | 1221 | ||
XM_017029446.1 | XP_016884935.1 | 13 | 1223 | ||
XM_017029447.1 | XP_016884936.1 | 13 | 1424 | ||
XM_017029448.1 | XP_016884937.1 | 13 | 1556 | ||
XM_017029449.1 | XP_016884938.1 | 12 | 1162 | ||
GenBank mRNA | AJ580093.1 | - | 12 | 1216 | |
AJ580094.1 | - | 12 | 1216 | ||
AK125986.1 | - | 2 | 204 | ||
BC132728.1 | - | 12 | 1152 | ||
BC132730.1 | - | 12 | 1152 |